Merck
  • Chromosome-wide aneuploidy study of cultured circulating myeloid progenitor cells from workers occupationally exposed to formaldehyde.
Chromosome-wide aneuploidy study of cultured circulating myeloid progenitor cells from workers occupationally exposed to formaldehyde.
Carcinogenesis (2014-11-14)
Qing Lan, Martyn T Smith, Xiaojiang Tang, Weihong Guo, Roel Vermeulen, Zhiying Ji, Wei Hu, Alan E Hubbard, Min Shen, Cliona M McHale, Chuangyi Qiu, Songwang Liu, Boris Reiss, Laura Beane-Freeman, Aaron Blair, Yichen Ge, Jun Xiong, Laiyu Li, Stephen M Rappaport, Hanlin Huang, Nathaniel Rothman, Luoping Zhang
RESUMO
Formaldehyde (FA) is an economically important industrial chemical to which millions of people worldwide are exposed environmentally and occupationally. Recently, the International Agency for Cancer Research concluded that there is sufficient evidence that FA causes leukemia, particularly myeloid leukemia. To evaluate the biological plausibility of this association, we employed a chromosome-wide aneuploidy study approach, which allows the evaluation of aneuploidy and structural chromosome aberrations (SCAs) of all 24 chromosomes simultaneously, to analyze cultured myeloid progenitor cells from 29 workers exposed to relatively high levels of FA and 23 unexposed controls. We found statistically significant increases in the frequencies of monosomy, trisomy, tetrasomy and SCAs of multiple chromosomes in exposed workers compared with controls, with particularly notable effects for monosomy 1 [P = 6.02E-06, incidence rate ratio (IRR) = 2.31], monosomy 5 (P = 9.01E-06; IRR = 2.24), monosomy 7 (P = 1.57E-05; IRR = 2.17), trisomy 5 (P = 1.98E-05; IRR = 3.40) and SCAs of chromosome 5 (P = 0.024; IRR = 4.15). The detection of increased levels of monosomy 7 and SCAs of chromosome 5 is particularly relevant as they are frequently observed in acute myeloid leukemia. Our findings provide further evidence that leukemia-related cytogenetic changes can occur in the circulating myeloid progenitor cells of healthy workers exposed to FA, which may be a potential mechanism underlying FA-induced leukemogenesis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Clorofórmio, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Clorofórmio, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Clorofórmio, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Clorofórmio, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Formaldeído, for molecular biology, 36.5-38% in H2O
SAFC
Formaldeído, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Clorofórmio, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Clorofórmio, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Clorofórmio, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Clorofórmio, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Formaldeído, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Clorofórmio, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Clorofórmio, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Clorofórmio, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Formaldeído, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Clorofórmio, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Formaldeído, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Clorofórmio, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Formaldeído, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Clorofórmio, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer