Documentos
Z282030
Coors alumina pestle
for Z281980
About This Item
Código UNSPSC:
41121800
NACRES:
NB.35
Materiais
alumina
embalagem
pack of 1 ea
fabricante/nome comercial
Coorstek 60356
pilão C
38 mm
Descrição geral
99.5% alumina. Extremely hard and completely nonabsorbent surface. Order mortar separately.
Informações legais
Coors is a trademark of Coorstek, Inc.
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 5
1 of 5
Agate mortar and pestle Standard form, O.D. 50 mm
Z136212
Agate mortar and pestle
Coors™ porcelain pestle for Z247480
Z247529
Coors porcelain pestle
Agate mortar and pestle Standard form, O.D. 65 mm
Z112518
Agate mortar and pestle
Agate mortar and pestle Standard form, O.D. 100 mm
Z168955
Agate mortar and pestle