Merck
22039
Swagelok® hintere Ferrule
Swagelok® 204-1, stainless steel, 1/8 in. Swagelok
Synonym(e):
Swagelok® 204-1,EDELSTAHL BACK FERRULE 1/8” PK5
About This Item
UNSPSC-Code:
41115717
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
stainless steel
Verpackung
pkg of 5 ea
Hersteller/Markenname
Swagelok® 204-1
Fitting
1/8 in. Swagelok
Allgemeine Beschreibung
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1]
Rechtliche Hinweise
Swagelok is a registered trademark of Swagelok Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)