Fortfahren mit
22028
Swagelok® hintere Ferrule
SWAGELOK® 404-1, stainless steel, 1/4 in. SWAGELOK
Synonym(e):
Swagelok® 404-1,SS BACK FERRULE 1/4” PK5
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41115717
NACRES:
SB.54
eCl@ss:
27150601
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
stainless steel
packaging
pkg of 5 ea
manufacturer/tradename
SWAGELOK® 404-1
technique(s)
gas chromatography (GC): suitable using :Suitable
fitting
1/4 in. SWAGELOK
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
220392202722069
description
SWAGELOK® 404-1, stainless steel, 1/4 in. SWAGELOK
description
SWAGELOK® 204-1, stainless steel, 1/8 in. SWAGELOK
description
SWAGELOK® 403-1, stainless steel, 1/4 in. SWAGELOK
description
PTFE back ferrule, SWAGELOK for 1/16 in. tubing
fitting
1/4 in. SWAGELOK
fitting
1/8 in. SWAGELOK
fitting
1/4 in. SWAGELOK
fitting
SWAGELOK for 1/16 in. tubing
packaging
pkg of 5 ea
packaging
pkg of 5 ea
packaging
pkg of 5 ea
packaging
pkg of 5 ea
material
stainless steel
material
stainless steel
material
stainless steel
material
PTFE back ferrule
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
manufacturer/tradename
SWAGELOK® 404-1
manufacturer/tradename
SWAGELOK® 204-1
manufacturer/tradename
SWAGELOK® 403-1
manufacturer/tradename
-
Still not finding the right product?
Explore all of our products under Swagelok® hintere Ferrule
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)