Merck
SYNF10401L
Synthware 47 mm filtration apparatus with fritted support
joint: ST/NS 40/35
Synonym(e):
Synthware F10401L, complete filtration apparatus
Größe auswählen
1 EA
€ 344,00
€ 344,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 344,00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
€ 344,00
Check Cart for Availability
Verbindung
joint: ST/NS40/35
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F10401L
Verwandte Kategorien
Allgemeine Beschreibung
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has a sealed-in coarse porosity fritted disc. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)