Merck
SYNF100003A
Synthware 47 mm filtration apparatus with stainless steel support
Synonym(e):
Synthware 47 mm filtration apparatus with stainless steel support, complete
Größe auswählen
1 EA
€ 160,00
€ 160,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 160,00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
€ 160,00
Check Cart for Availability
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F100003A
Allgemeine Beschreibung
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has removable 47 mm stainless steel support screen for use with viscous or proteinaceous solutions. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Ähnliches Produkt
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)