Merck
BR701364
BRAND® sealing film for microplates
self-adhesive, gas permeable, suitable for (for cell and tissue culture)
Synonym(e):
Accessories, PCR, Plates, Sealing films
Größe auswählen
100 EA
€ 103,00
€ 103,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 103,00
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.35
€ 103,00
Check Cart for Availability
Materialien
Rayon
Rayon film
Sterilität
non-sterile
Leistungsmerkmale
gas permeable
self-adhesive
Verpackung
pack of 100 ea
Hersteller/Markenname
BRAND 701364
Eignung
suitable for (for cell and tissue culture)
Allgemeine Beschreibung
When working with cell and tissue cultures, ensuring a secure seal is only the first step in protecting against contamination and facilitating safe transport. Equally critical is the ability to allow for proper gas exchange. Insufficient oxygen levels or excessive carbon dioxide can not only compromise the accuracy of experimental results but may also jeopardize cell viability. Consequently, films designed for cell and tissue cultures must meet stringent requirements to support both secure sealing and optimal gas exchange, ensuring a conducive environment for cellular growth and experimentation.
Leistungsmerkmale und Vorteile
  • Air permeable for optimum oxygen supply
  • Secure seal against contamination
  • Pierceability for easy sample extraction
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)