Skip to Content
Merck
All Photos(3)

Key Documents

N3393

Sigma-Aldrich

Naphthol Blue Black

BioReagent, suitable for electrophoresis

Synonym(s):

Acid Black 1, Amido Black 10B, Buffalo Black NBR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
Colour Index Number:
20470
Beilstein:
5374263
EC Number:
MDL number:
UNSPSC Code:
41105322
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

form

powder

composition

Dye content, ~85%

technique(s)

protein staining: suitable

impurities

≤10% water

suitability

suitable for electrophoresis

SMILES string

[Na+].[Na+].Nc1c(\N=N\c2ccc(cc2)[N+]([O-])=O)c(cc3cc(c(\N=N\c4ccccc4)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N6O9S2.2Na/c23-19-18-12(11-17(39(35,36)37)21(22(18)29)27-24-13-4-2-1-3-5-13)10-16(38(32,33)34)20(19)26-25-14-6-8-15(9-7-14)28(30)31;;/h1-11,29H,23H2,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b26-25+,27-24+;;

InChI key

AOMZHDJXSYHPKS-DROYEMJCSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Naphthol Blue Black has been used for the staining of western blot membrane for detection of all protein that are transferred to the membrane.

Biochem/physiol Actions

Naphthol blue black dye can be used to stain proteins on polyacrylamide gels, agarose gels and nitrocellulose membranes. After electrophoresis, fixing the proteins in the gel is recommended. The gel can then be stained with 0.1% Naphthol Blue Black in 7% (v/v) acetic acid for at least 2 hr and destained with a soluion of 7% (v/v) acetic acid. Detection sensitivity is approx. 20% that of Coomassie Blue R.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PopF1 and PopF2, two proteins secreted by the type III protein secretion system of Ralstonia solanacearum, are translocators belonging to the HrpF/NopX family.
Meyer D, et al.
Journal of Bacteriology, 188, 4903-4917 (2006)
Disc electrophoresis of proteins in the presence of sodium dodecyl sulphate.
E De Vito et al.
Experientia, 22(2), 124-125 (1966-02-15)
M D Chengalroyen et al.
World journal of microbiology & biotechnology, 29(4), 625-633 (2012-11-29)
Investigation into the biodegradative capability of several actinomycetes led to the discovery of many strains possessing an ability to decolorize a variety of azo and triphenylmethane dyes. Of particular interest is an Amycolatopsis sp. isolate which displayed the ability to
Shih-Hsien Chang et al.
Journal of hazardous materials, 166(2-3), 1279-1288 (2009-01-23)
Degradation of Remazol Brilliant Blue R (anthraquinone dye) and Acid Black 1 (azo dye) by Fenton oxidation and low-cost Fe(0)/air process was compared. The doses of Fenton reagent needed for decolorization of Remazol Brilliant Blue R was much higher than
Teresa M Lamb et al.
G3 (Bethesda, Md.), 3(12), 2273-2280 (2013-10-22)
Precise control of gene expression is a powerful method to elucidate biological function, and protein overexpression is an important tool for industry and biochemistry. Expression of the Neurospora crassa tcu-1 gene (NCU00830), encoding a high-affinity copper transporter, is tightly controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service