Skip to Content
Merck
All Photos(1)

Key Documents

A8181

Sigma-Aldrich

Amido Black Staining Solution 2X

electrophoresis reagent

Synonym(s):

Naphthol Blue Black solution, Amido Black 10B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
Beilstein:
5374263
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Quality Level

form

liquid

SMILES string

[Na+].[Na+].Nc1c(\N=N/c2ccc(cc2)[N+]([O-])=O)c(cc3cc(c(\C=C/c4ccccc4)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N6O9S2.2Na/c23-19-18-12(11-17(39(35,36)37)21(22(18)29)27-24-13-4-2-1-3-5-13)10-16(38(32,33)34)20(19)26-25-14-6-8-15(9-7-14)28(30)31;;/h1-11,29H,23H2,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b26-25+,27-24+;;

InChI key

AOMZHDJXSYHPKS-DROYEMJCSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Amido black (AB) is an acidic dye used for staining proteins. It can detect proteins levels of 50ng in both polyvinylidene difluoride (PVDF) and nitrocellulose membrane. AB aids protein visualization even at low concentrations. It has applications in forensics due to its ability to stain blood proteins in fingerprints.

Application

Amido Black Staining Solution 2X has been used in the staining:
  • serum proteins present in the culture supernatant
  • cysteine sulfinic acid decarboxylase protein in mouse liver homogenate
  • histones from chromatin samples

Reconstitution

After dilution with deionized water to a final volume of 500 ml, solution will contain 0.1% (w/v) amido black, 25% (v/v) isopropanol and 10% (v/v) acetic acid.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Placental release of taurine to both the maternal and fetal circulations in human term pregnancies
Holm MB, et al.
Amino Acids, 50(9), 1205-1214 (2018)
Detection of proteins on blot membranes
Goldman A, et al.
Current Protocols in Protein Science / Editorial Board, John E. Coligan ... [Et Al.], 86(1), 10-18 (2016)
Electrophoretic transfer of proteins from sodium dodecyl sulfate-polyacrylamide gels to a positively charged membrane filter.
J M Gershoni et al.
Analytical biochemistry, 124(2), 396-405 (1982-08-01)
The ATRX cDNA is prone to bacterial IS10 element insertions that alter its structure
Valle-Garcia D, et al.
SpringerPlus, 3(1), 222-222 (2014)
Meghan C Hughes et al.
The Journal of physiology, 598(7), 1377-1392 (2019-01-24)
Ninety-eight per cent of patients with Duchenne muscular dystrophy (DMD) develop cardiomyopathy, with 40% developing heart failure. While increased propensity for mitochondrial induction of cell death has been observed in left ventricle, it remains unknown whether this is linked to

Articles

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service