Skip to Content
Merck
All Photos(4)

Key Documents

C0625

Sigma-Aldrich

Caffeic acid

≥98.0% (HPLC)

Synonym(s):

3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
1954563
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98.0% (HPLC)

form

powder

mp

211-213 °C (dec.) (lit.)

solubility

ethanol: 50 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

OC(=O)\C=C\c1ccc(O)c(O)c1

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+

InChI key

QAIPRVGONGVQAS-DUXPYHPUSA-N

Gene Information

human ... ELA2(1991)
rat ... Alox5(25290)

Looking for similar products? Visit Product Comparison Guide

General description

Caffeic acid exists in its free and ester form. It is considered as the predominant polyphenol, contributing to the hydroxycinnamic acid content in various fruits. Coffee is one of the major source of caffeic acid.

Application

Caffeic acid has been used as a standard of phenolic acid in the study to determine the total phenolic acid content in vegetables after subjecting to alkaline and acid hydrolysis. It has also been used to determine its antioxidant activity by various assay methods.

Biochem/physiol Actions

A natural dietary phenolic compound found in plants that is an anti-oxidant. Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.
Anti-oxidant phenolic compound found in plants; Inhibits the synthesis of leukotrienes.
Caffeic acid and other polyphenols are metabolized by going through enzyme catalysed methyl transfer, sulfation and glucuronic acid addition. It non-competitively prevents the action of 5-lipoxygenase. Caffeic acid ready undergoes hydrolysis in alkaline conditions.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

related product

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparing antioxidative food additives and secondary plant products-use of different assays
Kranl K, et al.
Food Chemistry, 93(1), 171-175 (2005)
Caffeic acid is a selective inhibitor for leukotriene biosynthesis
Yasuko K, et al.
Biochimica et Biophysica Acta, Lipids and Lipid Metabolism, 792(1), 92-97 (1984)
Phenolic acids in potatoes, vegetables, and some of their products
Mattila P and Hellstrom J
J. Food Compos. Anal., 20(3-4), 152-160 (2007)
Polyphenols: food sources and bioavailability
Manach C, et al.
American Journal of Clinical Nutrition, 79(5), 727-747 (2004)
Javier Cifuentes et al.
Antioxidants (Basel, Switzerland), 10(7) (2021-07-03)
Non-centrifugal cane sugar (NCS) is a traditional sweetener in most sugarcane regions of the world. In Colombia, this product has a socio-economic importance due to the extensive cultivation area and the high consumption rate per capita. NCS traditional processing involves

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service