Skip to Content
Merck
All Photos(2)

Key Documents

306452

Sigma-Aldrich

Reactive Black 5

Dye content ≥50 %

Synonym(s):

Remazol Black B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21N5Na4O19S6
CAS Number:
Molecular Weight:
991.82
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, ≥50%

technique(s)

microbe id | staining: suitable

mp

>300 °C (lit.)

density

1.21 g/cm3 at 20 °C

λmax

597 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].Nc1c(N=Nc2ccc(cc2)S(=O)(=O)CCOS([O-])(=O)=O)c(cc3cc(c(N=Nc4ccc(cc4)S(=O)(=O)CCOS([O-])(=O)=O)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C26H25N5O19S6.4Na/c27-23-22-15(13-20(53(37,38)39)24(23)30-28-16-1-5-18(6-2-16)51(33,34)11-9-49-55(43,44)45)14-21(54(40,41)42)25(26(22)32)31-29-17-3-7-19(8-4-17)52(35,36)12-10-50-56(46,47)48;;;;/h1-8,13-14,32H,9-12,27H2,(H,37,38,39)(H,40,41,42)(H,43,44,45)(H,46,47,48);;;;/q;4*+1/p-4

InChI key

HFIYIRIMGZMCPC-UHFFFAOYSA-J

General description

Reactive Black 5 belongs to the class of textile azo dyes. It can be biodegraded by the laccase enzyme in Ganoderma lucidum.
Reactive Black 5 is a reactive dye belonging to vinyl sulphone type. Such reactive dyes serve as colorants for many industries including paper. The functional group of the dye forms a covalent link with substrate functional group.

Application

Reactive Black 5 (RB5) has been:
  • used in an optimization study on the photocatalytic degradation of RB5 dye in synthetic greywater effluent (i.e. aqueous RB5 in distilled water), in order to understand the effects of different photoreactor parameters on the degradation kinetics.
  • as a textile dye to investigate the anaerobic reduction in upflow stirred packed-bed reactors (USPBRs) with biological activated carbon (BAC) system
  • used in the performance evaluation of dye removal forlarge molecule substance

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Pérez et al.
Water environment research : a research publication of the Water Environment Federation, 85(4), 291-300 (2013-05-24)
In this research, ozonation of Reactive Black 5 (RB5) and Direct Red 28 (DR28) under the presence of Na2SO4 and Na2CO3 used as textile additives was investigated. The effect of these salts on discoloration, degradation dynamics, and the composition of
O Iglesias et al.
Environmental science and pollution research international, 20(4), 2172-2183 (2012-06-27)
The aim of this work was to improve the ability of the electro-Fenton process using Fe alginate gel beads for the remediation of wastewater contaminated with synthetic dyes and using a model diazo dye such as Reactive Black 5 (RB5).
Tailored activated carbons as catalysts in biodecolourisation of textile azo dyes
Mezohegyi G, et al.
Applied Catalysis. B, Environmental, 94(1-2), 179-185 (2010)
Irina Stambolova et al.
Journal of photochemistry and photobiology. B, Biology, 117, 19-26 (2012-09-29)
Spray pyrolysis procedure for preparation of nanostructured TiO(2) films with higher photocatalytic effectiveness and longer exploitation life is presented in this study. Thin films of active nanocrystalline TiO(2) were obtained from titanium isopropoxide, stabilized with acetyl acetone and characterized by
Deniz Uçar et al.
Water environment research : a research publication of the Water Environment Federation, 84(4), 323-327 (2012-07-28)
This study investigated the removal of Reactive Black 5 (RB5) textile dye from aqueous solutions using cotton (Gossypium hirsutum) seed shell (CSS) as low cost adsorbents. The data were described according to the Freundlich and Langmuir isotherm models. Of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service