Skip to Content
Merck
All Photos(2)

Key Documents

N9002

Sigma-Aldrich

Naphthol Blue Black

Dye content ~50 %, Powder

Synonym(s):

Acid Black 1, Amido Black 10B, Buffalo Black NBR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
Colour Index Number:
20470
Beilstein:
5374263
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Naphthol Blue Black, Dye content ~50 %

form

powder

Quality Level

composition

Dye content, ~50%

solubility

water: 1 mg/mL

λmax

617-620 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Nc1c(\N=N\c2ccc(cc2)[N+]([O-])=O)c(cc3cc(c(\N=N\c4ccccc4)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N6O9S2.2Na/c23-19-18-12(11-17(39(35,36)37)21(22(18)29)27-24-13-4-2-1-3-5-13)10-16(38(32,33)34)20(19)26-25-14-6-8-15(9-7-14)28(30)31;;/h1-11,29H,23H2,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b26-25+,27-24+;;

InChI key

AOMZHDJXSYHPKS-DROYEMJCSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Hsien Chang et al.
Journal of hazardous materials, 166(2-3), 1279-1288 (2009-01-23)
Degradation of Remazol Brilliant Blue R (anthraquinone dye) and Acid Black 1 (azo dye) by Fenton oxidation and low-cost Fe(0)/air process was compared. The doses of Fenton reagent needed for decolorization of Remazol Brilliant Blue R was much higher than
M D Chengalroyen et al.
World journal of microbiology & biotechnology, 29(4), 625-633 (2012-11-29)
Investigation into the biodegradative capability of several actinomycetes led to the discovery of many strains possessing an ability to decolorize a variety of azo and triphenylmethane dyes. Of particular interest is an Amycolatopsis sp. isolate which displayed the ability to
Amit Parikh et al.
Biotechnology letters, 27(5), 323-326 (2005-04-19)
Cyanobacterial cultures isolated from sites polluted by industrial textile effluents were screened for their ability to decolorize cyclic azo dyes. Gloeocapsa pleurocapsoides and Phormidium ceylanicum decolorized Acid Red 97 and FF Sky Blue dyes by more than 80% after 26
Gyorgy Csako
Methods in molecular biology (Clifton, N.J.), 869, 339-359 (2012-05-16)
Immunoelectrophoresis (IEP) was the first practical method that combined electrophoresis and -immunoprecipitation for identifying and characterizing proteins within complex mixtures. Over the years, IEP has been extended to include a variety of techniques and, as a general name, has been
Mary-Ann Glasier et al.
Current eye research, 33(8), 631-640 (2008-08-13)
To compare two variations of a membrane-based protein assay utilizing Amido black (AB) detection with a commercially available 3-(4-carboxybenzoyl) quinoline-2-carboxaldehyde (CBQCA) assay for use in the quantitation of individual tear proteins, pooled human tear proteins, and protein extracted from ex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service