Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Naphthol Blue Black
Synonym(s):
Acid Black 1, Amido Black 10B, Buffalo Black NBR
Empirical Formula (Hill Notation):
C
22
H
14
N
6
Na
2
O
9
S
2
CAS Number:
1064-48-8
Molecular Weight:
616.49
EC Number:
213-903-1
Products
Product Category
stains (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
N3393
Naphthol Blue Black,
BioReagent, suitable for electrophoresis
Expand
View Pricing
195243
Naphthol Blue Black,
Dye content 80 %
Expand
View Pricing
N9002
Naphthol Blue Black,
Dye content ~50 %, Powder
Expand
View Pricing