Skip to Content
Merck
All Photos(4)

Key Documents

E2129

Sigma-Aldrich

Evans Blue

Dye content ≥75 %, Powder

Synonym(s):

Direct Blue 53

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H24N6Na4O14S4
CAS Number:
Molecular Weight:
960.81
Colour Index Number:
23860
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Evans Blue, Dye content ≥75 %

form

powder

Quality Level

composition

Dye content, ≥75%

technique(s)

titration: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1\N=N\c2ccc3c(cc(c(N)c3c2O)S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(\N=N\c5ccc6c(cc(c(N)c6c5O)S([O-])(=O)=O)S([O-])(=O)=O)c(C)c4

InChI

1S/C34H28N6O14S4.4Na/c1-15-11-17(3-7-21(15)37-39-23-9-5-19-25(55(43,44)45)13-27(57(49,50)51)31(35)29(19)33(23)41)18-4-8-22(16(2)12-18)38-40-24-10-6-20-26(56(46,47)48)14-28(58(52,53)54)32(36)30(20)34(24)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4/b39-37+,40-38+;;;;

InChI key

ATNOAWAQFYGAOY-GPTZEZBUSA-J

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Evans blue (EB) C.I. 23860 is a synthetic bis-azo dye also known as T-1824 and Direct Blue 53. EB dye with higher water solubility has a strong affinity towards serum albumin, making it a high molecular weight protein tracer.

Application

Evans blue is a well-known vital stain with applications in histology and fluorescent microscopy. Evans blue has been used to check cell viability and study blood-brain barrier (BBB) permeability. Additional applications include the demonstration of specific pituitary cell types, selective staining of neutrophil plasma membranes, and fluorescence microscopy as a quenching agent to suppress unwanted autofluorescence.

Biochem/physiol Actions

Evans blue is a non-permeating dye. In presence of plasma membrane damage, the dye enters in the cytoplasm and nucleus, thereby staining them blue. Evans blue is used for checking cell viability.[1] It is also used to study BBB (blood-brain barrier) permeability. The dye binds to albumin and hence indicates BBB breakdown to protein.[2]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CO2 is the main inorganic C species entering photosynthetically active leaf protoplasts of the freshwater macrophyte Ranunculus penicillatus ssp. pseudofluitans.
Plant, Cell and Environment, 22, 1019-1026 (1999)
Peterson RL, et al.
Teaching Plant Anatomy Through Creative Laboratory Exercises (2008)
Yulian Zhou et al.
The Journal of clinical investigation, 124(9), 3825-3846 (2014-08-02)
Canonical WNT signaling is required for proper vascularization of the CNS during embryonic development. Here, we used mice with targeted mutations in genes encoding canonical WNT pathway members to evaluate the exact contribution of these components in CNS vascular development
Pilhan Kim et al.
Nature methods, 7(4), 303-305 (2010-03-17)
In vivo imaging of small animals offers several possibilities for studying normal and disease biology, but visualizing organs with single-cell resolution is challenging. We describe rotational side-view confocal endomicroscopy, which enables cellular imaging of gastrointestinal and respiratory tracts in mice
Brian P Eliceiri et al.
Methods in molecular biology (Clifton, N.J.), 686, 371-378 (2010-11-18)
The blood-brain barrier (BBB) is a monolayer of endothelial cells that is regulated by the proximity of a unique basement membrane and a tightly controlled molecular interaction between specialized subsets of cells including pericytes, astrocytes, and neurons. Working together, these

Questions

1–10 of 11 Questions  
  1. What amount  and concentration should I use Evans-Blaun injecting it into the ear skin of a mouse to see Evans-Blue in the draining Lymphnodes afterwards?

    1 answer
    1. This product has not been tested for any specific application, including in vivo use. Suitability would have to be determined experimentally by the end user. Historical information suggests that a concentration of 0.5% Evans Blue in PBS at pH 7.4 has been used successfully in staining tissue sections. This concentration may also be effective starting point for in vivo applications. Please refer to available literature for additional information.

      Helpful?

  2. How to prepare a stock 1% solution using E2129, and is ther any guidance on its use?

    1 answer
    1. A 0.5% solution of Evans Blue in PBS, pH 7.4. Product P3813 should be used as the PBS, which will serve as the stock solution. For use, 3-4 drops should be added to 100 mL of PBS P3813. This should be used as the final rinse with a 5-minute wash. After rinsing briefly, coverslip the sample.

      Helpful?

  3. What is the stability of the E2129 solution once prepared?

    1 answer
    1. The stability of the solution will depend on whether the solutions are sterile filtered or contaminated through routine use. It's recommended to store the solutions in the dark at 2-8°C.

      Helpful?

  4. Should Evans Blue solutions for fluorescent assays be aged or cured for 7 days before use?

    1 answer
    1. While some dyes require aging before use, there is no indication that Evans Blue solutions need to be aged or cured before use.

      Helpful?

  5. How do I dilute 80% purity evans blue powder into a 2% solution with normal saline?

    1 answer
    1. This product is soluble in water at 60 mg/mL. For typical laboratory staining applications, a 1X PBS solution is also commonly used. Solubility in normal saline has not been evaluated, but rates are expected to be similar. A 2% solution would be 20 mg/mL. The dye content is lot-specific and reported in the Certificate of Analysis. Please see the link below to access a sample or batch-specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/e2129#product-documentation

      Helpful?

  6. What is the solution stability of Product E2129, Evans Blue?

    1 answer
    1. This product is rather stable in aqueous solution and may be autoclaved at 15 psi for 30 min. However, dye made up in physiological saline should not be autoclaved.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. How should Product E2129, Evans Blue, be dissolved?

    1 answer
    1. This product is soluble in water, alcohol, acids and alkalies.

      Helpful?

  9. Can Evans Blue, Product E2129, be used as a counterstain for FITC?

    1 answer
    1. This product can be used as a counterstain in immunohistochemistry when using FITC. After staining for immunofluorescence, dip sections in a 0.1% (w/v) in water solution of Evans Blue for 5-10 minutes. Rinse well in fresh PBS or water before coverslipping. Reference: Immunocytochemistry, Theory and Practice, p. 82 (1988).

      Helpful?

  10. What is the shelf life of Product E2129, Evans Blue?

    1 answer
    1. This product has a retest date of 4 years.  This information can be found on the Certificate of Analysis.

      Helpful?

1–10 of 11 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service