Skip to Content
Merck
All Photos(4)

Documents

114413

Sigma-Aldrich

Bromothymol Blue

ACS reagent, Dye content 95 %

Synonym(s):

3′,3″-Dibromothymolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28Br2O5S
CAS Number:
Molecular Weight:
624.38
Beilstein:
373934
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

powder

composition

Dye content, 95%

color

pink-purple

visual transition interval

6.0-7.6, yellow to blue(passes test)

mp

200-202 °C ((392 - 396 °F) - lit.)
200-202 °C (lit.)

λmax

420 nm

ε (extinction coefficient)

≥17000 at 419-425 nm in methanol
≥8000 at 326-332 nm in methanol
≥9000 at 279-285 nm in methanol

storage temp.

room temp

SMILES string

CC(C)c1cc(c(C)c(Br)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c(O)c(Br)c4C

InChI

1S/C27H28Br2O5S/c1-13(2)17-11-20(15(5)23(28)25(17)30)27(19-9-7-8-10-22(19)35(32,33)34-27)21-12-18(14(3)4)26(31)24(29)16(21)6/h7-14,30-31H,1-6H3

InChI key

NUHCTOLBWMJMLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromothymol Blue, also known as, dibromothymolsulfonphthalein is a sulfonated hydroxyquinone in acidic aqueous solutions. In alkaline solutions, this dye is present as a moderately sized dianion. It is also known as bromthymol blue and dibromothymolsulfonphthalein. It is a pH-sensitive dye that changes color from yellow to blue.

Application

Bromothymol blue dye serves as a pH indicator because of the precise color transition at neutrality. Other applications include:
  • as a vital stain to trace the movement of fluids from the lymph and to define cell walls or nuclei under the microscope
  • for the demonstration of fungal hyphae within plant roots
  • in the assessment of the soluble acidity within single rice grains
  • use in an agar gel medium for enumeration of Bacillus cereus
  • as a component of several bacterial growth and detection substrates such as cystine lactose electrolyte deficient (CLED/BROLACIN); polymyxin pyruvate egg yolk mannitol bromothymol blue (PEMBA) and MacConkey agar.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andrea Pastore et al.
Talanta, 219, 121251-121251 (2020-09-06)
A pH colorimetric sensor array (CSA) with fast response time (<1 min) using only one acid-base indicator, Bromothymol Blue (BB), was prepared and characterized by modulating the amount, C, of the surfactant Hexadecyltrimethylammonium p-toluenesulfonate between 0 and 0.3725 gCTApTs/gprecursor with a
Reza Amin et al.
Micromachines, 11(6) (2020-07-01)
To transform from reactive to proactive healthcare, there is an increasing need for low-cost and portable assays to continuously perform health measurements. The paper-based analytical devices could be a potential fit for this need. To miniaturize the multiplex paper-based microfluidic
Mariana A Sanchez et al.
Analytica chimica acta, 694(1-2), 95-99 (2011-05-14)
Liquid-liquid microextraction without phase segmentation was implemented in a multicommuted flow system for determination of the anti-hypertensive diltiazem. The procedure was based on ion pair formation between the drug and the dye bromothymol blue at pH 3.5. The detection was
Bambang Kuswandi et al.
Talanta, 74(4), 613-618 (2008-03-29)
An optical fiber biosensor consisting of acetylcholinesterase (AChE) and bromothymol blue (BTB) doped sol-gel film was employed to detect organophosphate pesticide chlorpyrifos. The main advantage of this optical biosensor is the use of a single sol-gel film with immobilized AChE
Sevgi Tatar Ulu
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(3-4), 778-783 (2006-10-13)
A simple, rapid, accurate, precise and sensitive colorimetric method for the determination of finasteride in tablets is described. The proposed methods are based on the formation of ion-pair complexes between the examined drug with bromophenol blue (BPB), bromocresol green (BCG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service