Skip to Content
Merck
All Photos(1)

Key Documents

T57401

Sigma-Aldrich

Tridecane

≥99%

Synonym(s):

Cactus Normal Paraffin N 13, Cetiol iSAN, Paraffin N 13, n-Tridecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CH3
CAS Number:
Molecular Weight:
184.36
Beilstein:
1733089
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.4 (vs air)

vapor pressure

1 mmHg ( 59.4 °C)

Assay

≥99%

refractive index

n20/D 1.425 (lit.)

bp

110-112 °C/12 mmHg (lit.)
234 °C (lit.)

mp

−6-−4 °C (lit.)

density

0.756 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCC

InChI

1S/C13H28/c1-3-5-7-9-11-13-12-10-8-6-4-2/h3-13H2,1-2H3

InChI key

IIYFAKIEWZDVMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tridecane can be used as a:
  • Reagent for the hydrodechlorination of 4‑chlorophenol in the presence of bimetallic nanocrystals as a catalyst.
  • Reactant to synthesize α, ω-dicarboxylic acids through terminal oxygenation in the presence of an inorganic-biohybrid catalyst.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An unexpected mixture of substances in the defensive secretions of the tubuliferan thrips, Callococcithrips fuscipennis (Moulton)
Tschuch G, et al.
Journal of Chemical Ecology, 34(6), 742-747 (2008)
Chemical composition and biological activities of essential oil from the leaves of Sesuvium portulacastrum
Magwa ML, et al.
Journal of Ethnopharmacology, 103(1), 85-89 (2006)
Anil Kumar Mishra et al.
Journal of drug targeting, 12(9-10), 559-567 (2004-12-29)
The purpose of this study was to obtain the convenient, synthetically useful bifunctional chelating agent, 6-(4-isothiocyanatobenzyl)-5,7-dioxo-1,11-(carboxymethyl)-1,4,8,11-tetraazacyclotridecane, and to apply it to stable (99m)Tc-labelling of monoclonal antibodies (mAbs). The chelate was synthesised by reaction of nitrobenzyl malonate and triethylenetetramine followed by
Raúl A Laumann et al.
Journal of chemical ecology, 35(1), 8-19 (2009-01-08)
We tested the hypotheses that host-searching behavior of the egg parasitoids Telenomus podisi and Trissolcus basalis may be differentially influenced by the different blends of volatiles released from the metathoracic glands of adult stink bug host species. We further studied
A synthetic study of atropurpuran: construction of a pentacyclic framework by an intramolecular reverse-electron-demand Diels-Alder reaction.
Takahiro Suzuki et al.
Angewandte Chemie (International ed. in English), 50(39), 9177-9179 (2011-09-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service