Skip to Content
Merck
All Photos(3)

Key Documents

296821

Sigma-Aldrich

Nonane

anhydrous, ≥99%

Synonym(s):

n-Nonane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH3
CAS Number:
Molecular Weight:
128.26
Beilstein:
1696917
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
151 °C (lit.)
vapor pressure:
0.18 psi ( 37.7 °C)

grade

anhydrous

Quality Level

Agency

suitable for EPA 1613

vapor density

4.41 (vs air)

vapor pressure

0.18 psi ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

401 °F

expl. lim.

2.9 %

impurities

<0.005% water

evapn. residue

<0.0003%

refractive index

n20/D 1.405 (lit.)

bp

151 °C (lit.)

mp

−53 °C (lit.)

density

0.718 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC

InChI

1S/C9H20/c1-3-5-7-9-8-6-4-2/h3-9H2,1-2H3

InChI key

BKIMMITUMNQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonane is a linear alkane mainly found in petroleum-based fuels.

Application


  • Metabolomic analysis reveals Ligilactobacillus salivarius CCFM 1266 fermentation improves dairy product quality.: The research demonstrates the use of nonane as a volatile organic compound (VOC) marker in metabolomic studies to assess the quality improvement in dairy products through fermentation processes, enhancing food quality control (Yan et al., 2024).

  • Study on the effect of enzymolysis combined fermentation on reducing the off-flavor of egg white powder.: This paper discusses the application of nonane as an indicator for measuring the reduction of off-flavors in egg white powder, providing insights into improving the sensory attributes of food products (Chen et al., 2024).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation kinetics of n-nonane: Measurements and modeling of ignition delay times and product concentrations.
Rotavera B, et al.
Proceedings of the Combustion Institute, 33(1), 175-183 (2011)
Optimization of pressurized hot water extraction of Lavandin essential oils via central composite design.
Kamali H, et al.
International Journal of ChemTech Research, 6(11), 4853-4861 (2014)
Optimization of process variables for essential oil from Lavandula angustifolia by PHWE using central composite design.
Kamali H, et al.
International Journal of ChemTech Research, 6(2), 1151-1161 (2014)
Condensation of a supersaturated vapor. VIII. The homogeneous nucleation of n-nonane.
Hung CH, et al.
J. Chem. Phys. , 90(3), 1856-1865 (1989)
Johanna Faist et al.
European journal of medicinal chemistry, 45(1), 179-185 (2009-11-03)
N-Alkyl and N-(2-dialkylaminoethyl) derivatives of 5-amino-2-azabicyclo-nonanes were prepared and tested in vitro for their activities against the multidrug-resistant K1 strain of Plasmodium falciparum and Trypanosoma brucei rhodesiense (STIB 900). Most of the new compounds showed lower antitrypanosomal activity than their

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service