Skip to Content
Merck
All Photos(1)

Documents

87140

Sigma-Aldrich

Tetradecane

olefin free, ≥99.0% (GC)

Synonym(s):

n-Tetradecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH3
CAS Number:
Molecular Weight:
198.39
Beilstein:
1733859
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.83 (vs air)

vapor pressure

1 mmHg ( 76.4 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

455 °F

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

252-254 °C (lit.)

mp

5.5 °C (lit.)

density

0.762 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCC

InChI

1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3

InChI key

BGHCVCJVXZWKCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetradecane can be used for the synthesis of thermally stable nano-encapsulated phase change materials (NEPCMs), exhibiting thermal energy storage and heat transfer enhancement applications. It can also be used as an n-alkane model for the study of ignition time measurements for larger n-alkanes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A shock tube study of the ignition of n-heptane, n-decane, n-dodecane, and n-tetradecane at elevated pressures
Shen HPS, et al.
Energy and Fuels, 23(5), 2482-2489 (2009)
Preparation and characterization of nano-encapsulated n-tetradecane as phase change material for thermal energy storage
Fang G, et al.
Chemical Engineering Journal, 153(1-3), 217-221 (2009)
Vishwakarma Singh et al.
The Journal of organic chemistry, 74(16), 6092-6104 (2009-07-21)
A general and stereoselective methodology for the synthesis of bridged bicyclic octenones having various types of alkenyl chains and a tricyclic framework of secoatisanes and higher analogues is reported. In situ generation and cycloaddition of 2-allyl-6,6-spiroepoxycyclohexadienones with ethyl acrylate gave
Yong Zhou et al.
Journal of the American Chemical Society, 131(7), 2456-2457 (2009-02-10)
In this Communication, we report one-pot, high yield formation of unprecedented 1-D necklace-like chains consisting of hybrid Pd nanocrystals (NCs) embedded in spheres of a peptidic lipid. We employed the synthetic peptidic lipid, 2-(2-tetradecanamidoacetamido)acetic acid (1). We found that in
John J Beck et al.
Journal of agricultural and food chemistry, 57(9), 3749-3753 (2009-04-07)
Nonpareil almonds, Prunus dulcis , account for the largest percentage of almond varieties grown in the Central Valley of California. Several studies have investigated the various nonvolatile and volatile components of various plant parts; however, the volatile organic compound (VOC)

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service