Skip to Content
Merck
All Photos(2)

Key Documents

P3406

Sigma-Aldrich

Pentadecane

≥99%

Synonym(s):

n-Pentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH3
CAS Number:
Molecular Weight:
212.41
Beilstein:
1698194
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

7.4 (vs air)

Quality Level

vapor pressure

1 mmHg ( 91.6 °C)

Assay

≥99%

expl. lim.

6.5 %

refractive index

n20/D 1.431 (lit.)

bp

270 °C (lit.)

mp

8-10 °C (lit.)

density

0.769 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCC

InChI

1S/C15H32/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3-15H2,1-2H3

InChI key

YCOZIPAWZNQLMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentadecane is a linear alkane with high latent heat (~168 kJ/kg) and low melting point. These properties make it an ideal candidate as a phase change material (PCM) for cooling applications.[1][2][3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal properties of n-pentadecane, n-heptadecane and n-nonadecane in the solid/liquid phase change region.
Velez C, et al.
International Journal of Thermal Sciences, 94, 139-146 (2015)
The thermodynamic properties of n-pentadecane in the liquid state, determined by the results of measurements of sound velocity.
Dovnar D, et al.
High Temperature, 39(6), 835-839 (2001)
The capric?lauric acid and pentadecane combination as phase change material for cooling applications
Dimaano.RNM and Watanabe T
Applied Thermal Engineering, 22(4), 365-377 (2002)
Keith D Schwartz et al.
Organic letters, 13(2), 248-251 (2010-12-18)
Conjugate reduction of an enone accompanied by in situ intramolecular aldol condensation was used to construct the tetrasubstituted cyclohexane nucleus of phomactins. Subsequent relay ring-closing metathesis completed the nine-membered ansa bridge of the diterpenoid framework.
From 2,2'-methylenedifuran to all stereomeric pentadecane-1,3,5,7,9,11,13,15-octols.
M E Schwenter et al.
The Journal of organic chemistry, 66(23), 7869-7872 (2001-11-10)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service