Skip to Content
Merck
All Photos(1)

Documents

CRM47885

Supelco

Supelco 37 Component FAME Mix

certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in dichloromethane (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

shipped in

dry ice

storage temp.

-10 to -25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Check out our complete portfolio of Fatty acid methyl ester (FAME) reference material solution mixes

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl butyrate 400 μg/mL
    Methyl hexanoate 400 μg/mL
    Methyl octanoate 400 μg/mL
    Methyl decanoate 400 μg/mL
    Methyl undecanoate 200 μg/mL
    Methyl laurate 400 μg/mL
    Methyl tridecanoate 200 μg/mL
    Methyl myristate 400 μg/mL
    Methyl myristoleate 200 μg/mL
    Methyl pentadecanoate 200 μg/mL
    Methyl cis-10-pentadecenoate 200 μg/mL
    Methyl palmitate 600 μg/mL
    Methyl palmitoleate 200 μg/mL
    Methyl heptadecanoate 200 μg/mL
    cis-10-Heptadecenoic acid methyl ester 200 μg/mL
    Methyl stearate 400 μg/mL
    trans-9-Elaidic acid methyl ester 200 μg/mL
    cis-9-Oleic acid methyl ester 400 μg/mL
    Methyl linolelaidate 200 μg/mL
    Methyl linoleate 200 μg/mL
    Methyl arachidate 400 μg/mL
    Methyl γ-linolenate 200 μg/mL
    Methyl cis-11-eicosenoate ≤ 200 μg/mL
    Methyl linolenate 200 μg/mL
    Methyl heneicosanoate 200 μg/mL
    cis-11,14-Eicosadienoic acid methyl ester 200 μg/mL
    Methyl behenate 400 μg/mL
    cis-8,11,14-Eicosatrienoic acid methyl ester 200 μg/mL
    Methyl erucate 200 μg/mL
    cis-11,14,17-Eicosatrienoic acid methyl ester 200 μg/mL
    cis-5,8,11,14-Eicosatetraenoic acid methyl ester 200 μg/mL
    Methyl tricosanoate 200 μg/mL
    cis-13,16-Docosadienoic acid methyl ester 200 μg/mL
    Methyl lignocerate 400 μg/mL
    cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester 200 μg/mL
    Methyl nervonate 200 μg/mL
    cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 200 μg/mL
    See All (37)

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramesh Kumar Saini et al.
Frontiers in nutrition, 7, 578761-578761 (2020-11-27)
Precise information on the content and composition of nutritionally essential metabolites in food crops is a prerequisite for dietary recommendations and nutrient-dense food formulations. In the present study, the fatty acid profile of 21 Korean maize hybrids was analyzed by
Gunjan Singh et al.
3 Biotech, 8(10), 434-434 (2018-10-12)
This study investigated the potential of oleaginous yeast Rhodosporidium toruloides strain (ATCC20409) for the sustainable production of microbial lipids as biodiesel feedstock and other economically important fatty acids in comparison to algal or plant-based biodiesel. The strain exhibited high lipid
Mohsen Talebi et al.
Analytical and bioanalytical chemistry, 410(19), 4633-4643 (2017-12-08)
Twelve bis- or dicationic ionic liquids (ILs) including eight based on imidazolium, a single one based on phosphonium, and three based on pyrrolidinium cationic units were prepared with the bis(trifluoromethyl sulfonyl) imide anion. The two identical cationic moieties were attached
Helena M Amaro et al.
Marine drugs, 16(9) (2018-09-14)
The nutraceutical potential of microalgae boomed with the exploitation of new species and sustainable extraction systems of bioactive compounds. Thus, a laboratory-made continuous pressurized solvent extraction system (CPSE) was built to optimize the extraction of antioxidant compounds, such as carotenoids
Sanjeet Mehariya et al.
Antioxidants (Basel, Switzerland), 9(9) (2020-09-10)
The aim of this study was to develop an effective integrated cultivation system for Haematococcus pluvialis as a source of bioactive compounds such as astaxanthin, lutein, proteins, and fatty acids (FAs). The Chlorophyta H. pluvialis was cultivated in a vertical

Articles

This technical article will discuss GC Analysis of a 37-Component FAME Mix on Omegawax.

Protocols

GC Analyses of FAMEs by Boiling Point Elution

GC Analyses of FAMEs by Boiling Point Elution

GC Analyses of FAMEs by Boiling Point Elution

GC Analyses of FAMEs by Boiling Point Elution

See All

Chromatograms

suitable for GCsuitable for GCsuitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service