Skip to Content
Merck
All Photos(6)

Key Documents

183164

Sigma-Aldrich

(R)-(+)-Limonene

greener alternative

97%

Synonym(s):

(+)-p-Mentha-1,8-diene, (+)-Carvene, (R)-4-Isopropenyl-1-methyl-1-cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2204754
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.79

vapor density

4.7 (vs air)

Quality Level

vapor pressure

<3 mmHg ( 14.4 °C)

Assay

97%

form

liquid

optical purity

ee: 98% (GLC)

expl. lim.

6.1 %

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.473 (lit.)

bp

176-177 °C (lit.)

density

0.842 g/mL at 20 °C (lit.)

greener alternative category

SMILES string

CC(=C)[C@@H]1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m0/s1

InChI key

XMGQYMWWDOXHJM-JTQLQIEISA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Chiral intermediate for natural product synthesis.

Biochem/physiol Actions

Limonene is a cyclie terpene from Chinese medicinal herb essential oils used in the synthesis of carvone. Limonene may be used as a shrinking agent to dissolve polystyrene. Limonene may be used in various insecticidal and insect repellant applications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas G T Jaenson et al.
Journal of medical entomology, 43(1), 113-119 (2006-03-02)
In laboratory tests, ethyl acetate extracts of Hyptis suaveolens Poit. from Guinea-Bissau and Rhododendon tomentosum (Stokes) H. Harmaja (formerly Ledum palustre L.) and Myrica gale L. significantly reduced probing activity of Aedes aegypti (L.). In the field in southern Sweden
Jay M Patel et al.
Bio-protocol, 9(20), e3406-e3406 (2019-10-20)
Appetite is tightly linked to the sensory experience of feeding, including the smell, taste, and sight of food. Sensory perception can affect the palatability of food, modulating appetite beyond homeostatic requirements. Hypothalamic neurons that govern feeding are responsive to sensory
Fan Yang et al.
Frontiers in medicine, 8, 591830-591830 (2021-03-27)
At the time of the prevalence of coronavirus disease 2019 (COVID-19), pulmonary fibrosis (PF) related to COVID-19 has become the main sequela. However, the mechanism of PF related to COVID (COVID-PF) is unknown. This study aimed to explore the key
L Castillejos et al.
Journal of dairy science, 89(7), 2649-2658 (2006-06-15)
Two experiments were conducted to determine the effects of several essential oil active compounds on rumen microbial fermentation. In the first experiment, 4 doses (5, 50, 500, and 5,000 mg/L) of 5 essential oil compounds were evaluated using in vitro
Xiufeng Li et al.
ACS applied materials & interfaces, 12(13), 15802-15809 (2020-03-03)
Traditionally, complex coacervates of oppositely charged biopolymers have been used to form coatings around oil droplets for encapsulation of oil-soluble payloads. However, many proteins can form coacervates by themselves under certain conditions. Here, we revisit the well-known simple coacervates of

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service