Skip to Content
Merck
All Photos(1)

Documents

U407

Sigma-Aldrich

Undecane

≥99%

Synonym(s):

n-Undecane, Hendecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
Beilstein:
1697099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.4 (vs air)

Quality Level

vapor pressure

<0.4 mmHg ( 20 °C)

Assay

≥99%

refractive index

n20/D 1.417 (lit.)

bp

196 °C (lit.)

mp

−26 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCC

InChI

1S/C11H24/c1-3-5-7-9-11-10-8-6-4-2/h3-11H2,1-2H3

InChI key

RSJKGSCJYJTIGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Undecane is mainly used as a model n-alkane in studies relating to viscosities, excess molar enthalpies and vapor-liquid equilibrium of binary alkane mixtures.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A general correlation of vapor?liquid equilibria in hydrocarbon mixtures.
Chao K C and Seader J D
AIChE Journal, 7(4), 598-605 (1961)
Excess enthalpies of decan-1-ol+ n-pentane,+ n-hexane,+ n-heptane,+ n-octane,+ n-nonane,+ n-decane, and+ n-undecane at 298.15 K.
Kumaran M K and Benson G C
The Journal of Chemical Thermodynamics, 16(2), 175-182 (1984)
Measurements of the viscosity of n-heptane, n-nonane, and n-undecane at pressures up to 70 MPa.
Assael M J and Papadaki M
International Journal of Thermophysics, 12(5), 801-810 (1991)
Maya M Makatini et al.
Bioorganic & medicinal chemistry letters, 21(8), 2274-2277 (2011-03-25)
In this study, we present the first account of pentacycloundecane (PCU) peptide based HIV-protease inhibitors. The inhibitor exhibiting the highest activity made use of a natural HIV-protease substrate peptide sequence, that is, attached to the cage (PCU-EAIS). This compound showed
Igor Shamovsky et al.
Journal of medicinal chemistry, 52(23), 7706-7723 (2009-12-04)
The metabolic stability and selectivity of a series of CCR8 antagonists against binding to the hERG ion channel and cytochrome Cyp2D6 are studied by principal component analysis. It is demonstrated that an efficient way of increasing metabolic stability and selectivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service