Kluczowe dokumenty
58788
SSI Back-Pressure Regulator
About This Item
Kod UNSPSC:
41115700
Opis ogólny
Prevents bubbles and improves baseline stability. This 1/16 in. flange-type unit (4 × 1.3 cm) has a unique mechanism that ensures constant back-pressure over a wide range of mobile phase flows and viscosities. It is immune to clogging and other problems common to restrictor-type devices. Easily adjusts from 1-4 atm.(15-60 psi/1-4 kg/cm2).
Polecane produkty
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informacje prawne
SSI is a trademark of Scientific Systems, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)