Documenti
RAD195
radiello Clips
suspending Radiello triangular support plate, pk of 20
About This Item
Codice UNSPSC:
41104008
NACRES:
NB.24
impiego
suspending Radiello triangular support plate
Confezionamento
pk of 20
Produttore/marchio commerciale
Radiello
tecniche
passive air sampling: suitable
applicazioni
air monitoring
environmental
industrial hygiene
Note legali
radiello is a trademark of Institi Clinici Scientifici Maugeri
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti
Possiedi già questo prodotto?
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4
1 of 4
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Alberto Sturaro et al.
Environmental monitoring and assessment, 164(1-4), 93-100 (2009-04-04)
This paper reports the results of a long-term monitoring of benzene, toluene, ethylbenzene, xylenes (BTEX), phenol and cresols in the air of Padua during a wide period of the year 2007 using two radial passive samplers (Radiello system) equipped with
Joelle Roukos et al.
Environmental pollution (Barking, Essex : 1987), 157(11), 3001-3009 (2009-07-08)
Two measurement campaigns of volatile organic compounds (VOC) were carried out in the industrial city of Dunkerque, using Radiello passive samplers during winter (16-23 January) and summer (6-13 June) 2007. 174 compounds were identified belonging to six chemical families. Classifying
Articoli
Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocolli
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Contenuto correlato
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.