Documenti fondamentali
WHA77002110
Whatman® UNIFILTER® 384 well filter plates
clear polystyrene, non-sterile, DNA, 50 ea
Sinonimo/i:
Z746460, micro plate, multiwell plate, well plates
Scegli un formato
1 PKG
2.370,00 €
2.370,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 PKG
2.370,00 €
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.22
2.370,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Nome del prodotto
Whatman® UNIFILTER® plates 384, 384 well DNA binding plate, working volume 100 μL, filter bottom with long drip director
Materiali
conical bottom natural wells
natural polypropylene
polystyrene filter
Sterilità
non-sterile
Caratteristiche
filter bottom with long drip director
Confezionamento
pkg of 50 ea
Produttore/marchio commerciale
Cytiva
Whatman 7700-2110
Whatman Article No. 28420803 (US reference)
Volume massimo
100 μL
Misura
384 wells
Volume d’esercizio
100 μL
Tipo di binding
DNA high binding (membrane)
non-treated surface
Descrizione generale
Cytiva′s Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Polystyrene microplate for clean-up and cell capture
A high-throughput microplate can facilitate cell capture or dye removal and other sample clean-up processes requiring high recovery. Whatman UNIFILTER microplates are a good choice because of their long drip director and their ability to support gel filtration media.
Caratteristiche e vantaggi
  • Usable with gel filtration media, allowing high-throughput removal of dye blobs
  • Long drip director prevents well-to-well cross contamination
  • Rigid, clear polystyrene microplate can withstand centrifugation
  • Membrane media option of DNA binding polymer or glass microfiber
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)