DWK885300-0001
KIMBLE® Dounce tissue grinder set
capacity (1 mL)
Sinonimo/i:
Tissue Grinder
Scegli un formato
203,00 €
Scegli un formato
About This Item
203,00 €
Prodotti consigliati
Materiali
borosilicate glass
Confezionamento
case of 1 ea
Produttore/marchio commerciale
Kimble® 885300-0001
Capacità
(1 mL)
Descrizione generale
- All-glass
- Two pestles are supplied with each complete unit
- Large clearance pestle is used for the initial sample reduction
- Small clearance pestle is used to form the final homogenate
- Replacement components are available and completly interchangeable
- Manufactured from 33 expansion, low extractaable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Caratteristiche e vantaggi
- Manufactured from borosilicate glass 3.3
- Designed primarily for cellular work where the nucleus remains intact after homogenization.
- All-glass construction
- Two pestles are supplied with each complete unit
- Large clearance pestle is used for the initial sample reduction
- Small clearance pestle is used to form the final homogenate
- Replacement components are available and completely interchangeable
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)