Documenti fondamentali
DWK885300-0000
KIMBLE® Dounce tissue grinder set
capacity (0.5 mL)
Sinonimo/i:
KIMBLE tissue grinder, Tissue homogenizer
Scegli un formato
1 EA
203,00 €
203,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
203,00 €
About This Item
Codice UNSPSC:
41101703
NACRES:
NB.74
203,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
borosilicate glass
Confezionamento
case of 1 ea
Produttore/marchio commerciale
Kimble® 885300-0000
Capacità
(0.5 mL)
Descrizione generale
Dounce All-Glass tissue grinders designed for cellular work in which the nucleus remains intact after homogenization
  • All-glass
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completly interchangeable
  • Manufactured from 33 expansion, low extractaable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Caratteristiche e vantaggi
  • Manufactured from borosilicate glass 3.3
  • Designed primarily for cellular work where the nucleus remains intact after homogenization.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)