Documenti fondamentali
CLS3294050M
Pyrex® Gooch crucible, high form, with fritted disc
capacity 50 mL, porosity 10-15 μm
Sinonimo/i:
Corning® 32940
About This Item
Codice UNSPSC:
41121812
Il prodotto CLS3294050M al momento non è in vendita nel tuo paese Contatta l'Assistenza Tecnica.
Produttore/marchio commerciale
Corning 32940-50M
Capacità
50 mL
Diam. disco
40 mm
Dimensione pori
10-15 μm porosity
Descrizione generale
Particularly adapted to analytical work where precipitates are dried to constant weight at 110°C. Also suitable for higher temperatures, in which case is advisable to heat in an electric furnace.
Crucibles should not be subjected to sudden temperature changes.
In order to avoid strain, they should not be removed from the furnace until the temperature has dropped to 250°C.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)