BR458734
BRAND® filter crucible
borosilicate glass, capacity 50 mL, 2 D 4
Sinonimo/i:
Filter, Filter crucible
About This Item
Codice UNSPSC:
41122200
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
Produttore/marchio commerciale
BRAND 458734
Capacità
50 mL
Dimensione pori
10-16 μm
Descrizione generale
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)