Ugrás a tartalomra
Merck
  • Evaluation of Antibacterial and Cytotoxicity Properties of Silver Nanowires and Their Composites with Carbon Nanotubes for Biomedical Applications.
Evaluation of Antibacterial and Cytotoxicity Properties of Silver Nanowires and Their Composites with Carbon Nanotubes for Biomedical Applications.
International journal of molecular sciences (2020-04-01)
Arianna De Mori, Richard S Jones, Matteo Cretella, Guido Cerri, Roger R Draheim, Eugen Barbu, Gianluca Tozzi, Marta Roldo
KIVONAT
In this work, we prepared silver nanowires (AgNWs) via the polyol method in the presence or absence of single wall carbon nanotubes (CNTs) and tested their physicochemical, antibacterial and cytotoxic properties. Results showed that the introduction of CNTs lead to the formation of AgNWs at lower temperature, but the final product characteristics of AgNWs and AgNWs-CNT were not significantly different. AgNWs exhibited antibacterial properties against all the studied bacterial species via the formation of oxygen reactive species (ROS) and membrane damage. Furthermore, AgNWs exhibited a dose-dependent and time-dependent toxicity at concentrations ≥ 10 µg/mL. Fibroblasts appeared to be more resistant than human colorectal adenocarcinoma (Caco-2) and osteoblasts to the toxicity of AgNWs. The cytotoxicity of AgNWs was found to be related to the formation of ROS, but not to membrane damage. Overall, these results suggest that AgNWs are potential antibacterial agents against E. coli, S. aureus, MRSA and S. saprophyticus, but their dosage needs to be adjusted according to the route of administration.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Octanal, 99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DL-Lactic acid, ~90% (T)