Fontos dokumentumok
Z719498
Sorenson low binding Ultra G microtips
maximum volume 10 μL, pack of 1000 ea (Bag), non-sterile
About This Item
UNSPSC kód:
41121600
NACRES:
NB.01
material
colorless
polypropylene
sterilitás
non-sterile
Jellemzők
barrier: no
graduations
kiszerelés
pack of 1000 ea (Bag)
gyártó/kereskedő neve
Sorenson 23580T
maximum térfogat
10 μL
Általános leírás
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for ultra-micro pipettors.
Jogi információk
Sorenson is a trademark of Sorenson BioScience, Inc.