Fontos dokumentumok
EP5820770007
Adapter for Eppendorf® FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors
capacity, 1 HPLC tube, pack of 4
About This Item
UNSPSC kód:
41103911
NACRES:
NB.82
Jellemzők
capacity 1 HPLC tube
kiszerelés
pack of 4
gyártó/kereskedő neve
Eppendorf® 5820770007 (Global)
technika/technikák
centrifugation: suitable
kompatibilitás
for use with FA-45-16-17
for use with FA-45-20-17
for use with Rotor FA-45-12-17
Jogi információk
Eppendorf is a registered trademark of Eppendorf AG