Fontos dokumentumok
EP5353040113
Eppendorf® Tube holder PCR 96
Holds 96 x 0.2 mL PCR-tubes, -strips or 96-well PCR-plates, semi- skirted or unskirted
About This Item
UNSPSC kód:
41121800
NACRES:
NC.21
Jellemzők
Holds 96 x 0.2 mL PCR-tubes, -strips or 96-well PCR-plates, semi- skirted or unskirted
gyártó/kereskedő neve
Eppendorf® 5353040113
technika/technikák
PCR: suitable
Jogi információk
Eppendorf is a registered trademark of Eppendorf AG