Merck
Z763489
MAGic Clamp magnetic clamp
holds 50 mL Erlenmeyer flasks (max. 20)
Synonym(e):
flask clamps, magnetic clamps
Größe auswählen
1 EA
42,30 €
42,30 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
42,30 €
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.77
42,30 €
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific H1000-MR-50
Allgemeine Beschreibung
The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)