Merck
Z763535
MAGic Clamp magnetic clamp
holds 2000 mL Erlenmeyer flasks (max. 2)
Synonym(e):
flask clamps, magicclamp, magnetic clamps
Größe auswählen
1 EA
100,00 €
100,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
100,00 €
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.77
100,00 €
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific H1000-MR-2000
Allgemeine Beschreibung
The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)