Merck
Z763403
Orbi-Shaker XL orbital shaker with flat mat platform
AC/DC input 230 V AC, UK plug
Synonym(e):
orbital mixers, orbital shakers, stirring plates
Größe auswählen
1 EA
3.810,00 €
3.810,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
3.810,00 €
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.77
3.810,00 €
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific BT1011-E-UK
Parameter
30-300 rpm speed
Gesamt L × B × H
18.5 in. (47 cm) × 21.25 in. (54 cm) × 5 in. (12.5 cm)
Plattform L × B
17.8 in. (45 cm) × 17.8 in. (45 cm)
Gewicht
30 kg
Allgemeine Beschreibung
The Orbi-Shaker XL offers flask capacity that far exceeds most bench-top models. With the large platform and powerful, beltless motor drive, the Orbi-Shaker XL is capable of mixing up to 5 x 2 L flasks or 9 x 1 L flasks simultaneously. The included non-slip rubber mat platform is ideal for low speed applications, while the convenient MAGic Clamp platform is also available for high speed mixing of Erlenmeyer flasks, tube racks or microplates. The MAGic Clamp allows for instant flask clamp exchange without the use of tools, screws or any other hardware.
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Orbi-Shaker is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)