Key Documents
Z227250
Supelco® PEEK Fitting
One-piece fingertight, pkg of 5 ea
Synonyma:
PEEK Fitting
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
packaging
pkg of 5 ea
General description
Biocompatible fittings for all 1/16 in. tubing connections.
Features and Benefits
Holds to 6000 psi (420 kg/cm2).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
Je nám líto, ale pro tento produkt momentálně nemáme COA k dispozici online.
Zákazníci si také prohlíželi
Slide 1 of 1
1 of 1
Upchurch Fittings Kit for use with Waters
58639
Upchurch Fittings Kit