Merck
57692
RheFlex® PEEK Fittings
Ferrule pack, 1/16 in. tubing
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
UNSPSC Code:
41115700
NACRES:
NB.21
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
packaging
pkg of 5 ea
compatibility
1/16 in. tubing
General description
PEEK (polyetheretherketone) fingertight fittings (ferrule pack) are convenient, inert, and biocompatible. Use these fittings with 1/16 in. O.D. PEEK, stainless steel, titanium, Tefzel, or PTFE tubing. PEEK fittings are compatible with all HPLC solvents (avoid concentrated sulfuric and nitric acids), and can be used at temperatures up to 150° C. Unlike stainless steel ferrules, PEEK ferrules do not permanently lock into place on the tubing. This allows the interchanging of fittings with tubing and columns, from manufacturer to manufacturer (e.g. from Agilent to Waters), and still form a zero dead volume, leak-free connection.
Features and Benefits
For Rheodyne valves. Hold to 5000 psi (350 kg/cm²).
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
RheFlex is a registered trademark of IDEX Health & Science, LLC
Still not finding the right product?
Explore all of our products under RheFlex® PEEK Fittings
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)