Key Documents
22749
Supelco
Fritted Purge Sampler
size 25 mL, top stem, Tekmar® 14-4007-024
About This Item
UNSPSC Code:
41115700
material
glass
Quality Level
agency
EPA 603
manufacturer/tradename
Tekmar® 14-4007-024
size
25 mL
application(s)
industrial hygiene
Features and Benefits
Note: All 1/4 in. O.D. glass ends are ground glass.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)