Hlavní dokumenty
22789
Supelco
Fritted Purge Sampler
size 25 mL, Tekmar® 14-2334-024
About This Item
UNSPSC Code:
41115700
material
glass
Quality Level
manufacturer/tradename
Tekmar® 14-2334-024
size
25 mL
application(s)
industrial hygiene
compatibility
for use with Tekmar® 2000/3000
for use with standard aqueous analysis
Features and Benefits
Note: All 1/4 in. O.D. glass ends are ground glass.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)