Key Documents
EP0030119851
Eppendorf® Cuvette Rack
Holds 30 x cuvettes, 3 rows of 10 wells each, pkg of 2 pieces, white, Autoclavableautoclavable
About This Item
UNSPSC Code:
41122800
NACRES:
NB.11
material
polypropylene
feature
3 rows of 10 wells each
Autoclavable
autoclavable
Holds 30 x cuvettes
packaging
pkg of 2 pieces
manufacturer/tradename
Eppendorf® 30119851
color
white
Legal Information
Eppendorf is a registered trademark of Eppendorf AG