Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
2 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 300,00 Kč |
1 pkg | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 300,00 Kč |
O této položce
1 300,00 Kč
material
polypropylene , white
feature
autoclavable, Holds 12 x 5-15 mL tubes
packaging
pkg of 2 pieces
manufacturer/tradename
Eppendorf® 30119835
color
white
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description Holds 12 x 5-15 mL tubes, pkg of 2 pieces, white, autoclavable | description Holds 48 x 0.5 mL tubes, 3 rows of 16 wells each, pkg of 2 pieces, white, autoclavable | description Holds 36 x 1.5-2 mL tubes, 3 rows of 12 wells each, pkg of 2 pieces, white, autoclavable | description Holds 30 x cuvettes, 3 rows of 10 wells each, pkg of 2 pieces, white, Autoclavableautoclavable |
| manufacturer/tradename Eppendorf® 30119835 | manufacturer/tradename Eppendorf® 30119800 | manufacturer/tradename Eppendorf® 30119819 | manufacturer/tradename Eppendorf® 30119851 |
| feature Holds 12 x 5-15 mL tubes, autoclavable | feature autoclavable, 3 rows of 16 wells each, Holds 48 x 0.5 mL tubes | feature Holds 36 x 1.5-2 mL tubes, 3 rows of 12 wells each, autoclavable | feature Autoclavable |
| material polypropylene , white | material polypropylene | material polypropylene | material polypropylene |
| packaging pkg of 2 pieces | packaging pkg of 2 pieces | packaging pkg of 2 pieces | packaging pkg of 2 pieces |
| color white | color white | color white | color white |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?