Hlavní dokumenty
HS29025G
Microtube Rack
assorted colors rack
Synonyma:
Microcentrifuge Tube Holder, Microtube Storage Rack
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
material
assorted colors rack
polypropylene
feature
to hold 80 tube(s)
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS29025G
L × W × H
225 mm × 67 mm × 28 mm
General description
The 80-Well Microtube Rack is a popular fraction-collector-plate style rack.
Features and Benefits
  • Easily fill and transfer 80 tubes in compact arrangement (5x16 array)
  • Frosted labeling area on bottom of rack
  • Embossed Index Numbers for quick sample ID and retrieval
  • Made of polypropylene
  • Autoclavable
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)