Hlavní dokumenty
EP0030119800
Eppendorf® Tube Rack for 0.5 mL Tubes
Holds 48 x 0.5 mL tubes, 3 rows of 16 wells each, pkg of 2 pieces, white, autoclavable
About This Item
UNSPSC Code:
41122800
NACRES:
NB.11
material
polypropylene
feature
autoclavable
3 rows of 16 wells each
Holds 48 x 0.5 mL tubes
packaging
pkg of 2 pieces
manufacturer/tradename
Eppendorf® 30119800
color
white
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)