Key Documents
BAF441710000
Microscope View-Pack®
Replacement Slide Holder Pages, pack of 10 sheets
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
form
sheet
feature
16 pockets per page
packaging
pack of 10 sheets
manufacturer/tradename
Bel-Art F44171-0000
slot size
1 × 3 in.
General description
The standard 23 x 30cm (9" x 12") 3-ring loose-leaf binder includes ten 22 x 27cm (8 1/2 " x 10 1/2 ") vinyl View-Pack Microscope Slide Holder Pages (sufficient for 160 slides). Every page has pockets featuring a white back and transparent front. A center flap holds slides securely. Folding the page along the middle crease allows slides to be conveniently retrieved. View-Pack Microscope Slide Holder Pages may also be purchased separately.
Organize, display and transport microscope slides
  • Includes ten 8 1/2 " x 10 1/2 " vinyl microscope slide holder pages
  • 16 slides 25 x 75mm (1" x 3") are held in separate pockets on every page
Legal Information
View-Pack is a registered trademark of SP Scienceware, Division of SP