Hlavní dokumenty
HS15981B
Slide holder
for 78 slides, white polyethylene
About This Item
UNSPSC Code:
41123302
NACRES:
NB.35
material
white polyethylene
feature
Holds 78 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15981B
size
299x178x21 mm
General description
The Slide Holder holds slides during evaluation, cleaning, and drying.
  • Made from chemical-resistant polyethylene
  • Solid, strong construction with rubber feet for bench top stability
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)