Merck
R1145
SAFC
Medio RPMI-1640
Without ʟ-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture, 10x
Fabricación farmacéutica
Sinónimos:
RPMI Medium, RPMI-1640 Culture Medium, Roswell Park Memorial Institute 1640 medium
About This Item
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
product name
Medio RPMI-1640, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Quality Level
sterility
sterile-filtered
form
liquid
concentration
10 ×
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
NaHCO3: no
phenol red: yes
sodium pyruvate: no
HEPES: no
L-glutamine: no
shipped in
ambient
storage temp.
2-8°C
General description
El medio RPMI 1640 lo desarrolló el equipo de Moore en el Instituto Roswell Park Memorial en 1966. Se formuló una modificación del medio McCoy 5A para sostener las células linfoblastoides en cultivo en suspensión, pero desde entonces se ha demostrado que sirve de soporte para una amplia variedad de células que necesitan anclaje. Originalmente indicado para utilizar con un complemento sérico, se ha demostrado que el RPMI 1640 respalda diversas líneas celulares en ausencia de suero. También se ha utilizado ampliamente en protocolos de fusión y en el crecimiento de células híbridas. Este medio es adecuado para el cultivo de leucocitos humanos sanos y neoplásicos.
Reconstitution
Supplement with 0.3 g/L L-glutamine, 2.0 g/L sodium bicarbonate, 0.001 g/L folic acid at 1×.
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Visite la Librería de documentos
Ryoichi Arita et al.
Diabetes, 58(1), 215-226 (2008-10-09)
Leukocyte adhesion in retinal microvasuculature substantially contributes to diabetic retinopathy. Involvement of the Rho/Rho kinase (ROCK) pathway in diabetic microvasculopathy and therapeutic potential of fasudil, a selective ROCK inhibitor, are investigated. Localization of RhoA/ROCK and Rho activity were examined in
Fhernanda R Smiderle et al.
BMC complementary and alternative medicine, 11, 58-58 (2011-07-27)
Mushroom polysaccharides have traditionally been used for the prevention and treatment of a multitude of disorders like infectious illnesses, cancers and various autoimmune diseases. Crude mushroom extracts have been tested without detailed chemical analyses of its polysaccharide content. For the
Tamara Veiga-Parga et al.
PLoS pathogens, 7(12), e1002427-e1002427 (2011-12-17)
Ocular herpes simplex virus infection can cause a blinding CD4⁺ T cell orchestrated immuno-inflammatory lesion in the cornea called Stromal Keratitis (SK). A key to controlling the severity of SK lesions is to suppress the activity of T cells that
Yoichi Matsuo et al.
International journal of cancer, 124(4), 853-861 (2008-11-28)
CXC-chemokines are involved in the chemotaxis of neutrophils, lymphocytes and monocytes. However, role of these chemokines in tumorigenesis, especially with regard to interaction between tumor and its microenvironment, has not been clearly elucidated. The purpose of this study was to
David-Alexandre Trégouet et al.
Diabetes, 57(10), 2843-2850 (2008-07-18)
Genetic and environmental factors modulate the susceptibility to diabetic nephropathy, as initiating and/or progression factors. The objective of the European Rational Approach for the Genetics of Diabetic Complications (EURAGEDIC) study is to identify nephropathy susceptibility genes. We report molecular genetic