Merck
MTCM4025
MTC Bio Flexible Inoculating Loops
capacity 10 μL, sterile, pack of 1000 ea (50 x bags 20 ea)
Synonym(e):
culture needles
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.15
Sterilität
sterile
Verpackung
pack of 1000 ea (50 x bags 20 ea)
Hersteller/Markenname
MTC Bio M4025
Kapazität
10 μL
Allgemeine Beschreibung
Inoculating loops, 10ul, flexible PP, 50 sterile peel bags of 20 loops, 1000/pk
Leistungsmerkmale und Vorteile
  • Sterile and disposable, no flaming
  • Uniform stroking and inoculating
  • Hexagonal grip - improves control
  • Smooth loop surface - no gouging
Rechtliche Hinweise
MTC is a trademark of MTC Bio