Merck
C3983
Sigma-Aldrich
Scienceware® cloning cylinders, polystyrene
I.D. × H 6.4 mm × 8 mm
Synonym(e):
cloning cylinder, cloning equipment
Größe auswählen
50 EA
CHF 111.00
CHF 111.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
50 EA
CHF 111.00
About This Item
UNSPSC-Code:
41106618
NACRES:
NB.22
CHF 111.00
Check Cart for Availability
Sterilität
sterile
Verpackung
pack of 50 ea
Hersteller/Markenname
Bel-Art F37847-0200
ID × H
6.4 mm × 8 mm
Allgemeine Beschreibung
To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.
Rechtliche Hinweise
Scienceware is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)