Merck
396966
Sigma-Aldrich
Decamethoniumbromid
Ion-Pair reagent, ≥99%
Synonym(e):
Decamethylen-bis(trimethylammoniumbromid), Decan-1,10-bis-trimethylammoniumbromid
About This Item
Lineare Formel:
(CH3)3N(Br)(CH2)10N(Br)(CH3)3
CAS-Nummer:
Molekulargewicht:
418.29
Beilstein:
3728288
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352103
PubChem Substanz-ID:
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Qualität
Ion-Pair reagent
Qualitätsniveau
Assay
≥99%
mp (Schmelzpunkt)
263-267 °C (lit.)
SMILES String
[Br-].[Br-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C
InChI
1S/C16H38N2.2BrH/c1-17(2,3)15-13-11-9-7-8-10-12-14-16-18(4,5)6;;/h7-16H2,1-6H3;2*1H/q+2;;/p-2
InChIKey
HLXQFVXURMXRPU-UHFFFAOYSA-L
Piktogramme
Skull and crossbones
Signalwort
Danger
Gefahreneinstufungen
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Lagerklassenschlüssel
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Dana B Finley et al.
Journal of pharmacological and toxicological methods, 59(2), 108-119 (2009-04-16)
Exposure to irreversible cholinesterase (ChE)-inhibiting compounds, such as organophosphates may produce neuromuscular dysfunction. However, less is known about changes in neuromuscular transmission after treatment with reversible ChE-inhibitors. These studies adapted single fiber electromyography (SFEMG) techniques to quantify neuromuscular jitter in
Chigusa Sawamura et al.
Magnetic resonance in medicine, 56(6), 1235-1241 (2006-11-08)
To examine the effect of immobilization on the development of articular cartilage, we assessed glycosaminoglycan (GAG) content in the chick articular surface by delayed gadolinium-enhanced MRI of cartilage (dGEMRIC). Chick embryos were paralyzed by decamethonium bromide (DMB) from day 10
J A Germiller et al.
Journal of pediatric orthopedics, 18(3), 314-318 (1998-05-26)
Clubfoot is a birth defect that may be related to muscle weakness or imbalance. The purpose of this study was to examine the relationships among muscle and tendon size and embryonic motility in a paralyzed chick embryo model of clubfoot
Roger L Papke et al.
The Journal of pharmacology and experimental therapeutics, 333(2), 501-518 (2010-01-27)
Transgenic mouse models with nicotinic acetylcholine receptor (nAChR) knockouts and knockins have provided important insights into the molecular substrates of addiction and disease. However, most studies of heterologously expressed neuronal nAChR have used clones obtained from other species, usually human
Raymond Macharia et al.
Anatomy and embryology, 208(1), 75-85 (2004-03-05)
This study determined the effect of decamethonium bromide (DMBr), a non-competitive blocker of the neuromuscular junction, on skeletal muscle development during chick embryogenesis. Decamethonium bromide caused generalized edema and high mortality with treated embryos rarely surviving beyond day 16 of